An improved process of isomaltooligosaccharide production in kimchi involving the addition of a Leuconostoc starter and sugars.
International journal of food microbiology (2013-12-03)
Seung Kee Cho, Hyun-Ju Eom, Jin Seok Moon, Sae-Bom Lim, Yong Kook Kim, Ki Won Lee, Nam Soo Han
RESUMEN
Isomaltooligosaccharides (IMOs) are α-(1→6)-linked oligodextrans that show a prebiotic effect on Bifidobacterium spp. This study sought to improve IMO synthesis during lactate fermentation in kimchi by inoculating the kimchi fermentation mix with a starter and sugars; the psychrotrophic Leuconostoc citreum KACC 91035 strain with high dextransucrase activity was used as a starter and sucrose (58 mM) and maltose (56 mM) were added as the donor and acceptor for the glucose-transferring reaction of the dextransucrase, respectively. With the addition of both the starter and the sugars and incubation at 10°C, IMOs were produced in kimchi after 3d. Without the starter, the IMO production rate and maximal concentration in kimchi were 15.05 mM/d and 75.27 mM, respectively, whereas with the starter, the rate and concentration increased to 22.04 mM/d and 110.19 mM, respectively. In addition, the sucrose-maltose mix gave an appropriate level of sweetness by releasing fructose and prevented unfavorable polymer synthesis by IMO production. This result suggests that lactic acid bacteria expressing a highly active glycosyltransferase can be used for the synthesis of beneficial oligosaccharides in various fermented foods.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Supelco
Sucrose, analytical standard
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20