Merck
Influence of water stress on antioxidative enzymes and yield of banana cultivars and hybrids.
Pakistan journal of biological sciences : PJBS (2014-02-13)
K Krishna Surendar, D Durga Devi, I Ravi, P Jeyakumar, K Velayudham
RESUMEN
The main aim for this field experimental study is to screening of various banana cultivars and hybrids for water deficit tolerance through antioxidative enzymes and yield. The field experiment was conducted at National Research Centre for Banana to screen the banana cultivars and hybrids for water deficit tolerance and to elucidate information on antioxidative enzymes mechanism correlated with yield of banana cultivars and hybrids. The methodology of this experimental were analyzed by split plot design and has two treatments considered as main plot (M) viz., control plot taken as M1 and water deficit plot taken as M2 and also the cultivars and hybrids were considered as sup plots (S). The stress was imposed according to the Available Soil Moisture (ASM) and this ASM was measured by using pressure plate membrane apparatus. The experimental data were significantly varied between the treatments and also cultivars and hybrids. The antioxidative enzymes of catalase, super oxide dismutase and ascorbate peroxidase were significantly enhanced during water deficit conditions. Among the twelve cultivars and hybrids, Karpuravalli, Karpuravalli x Pisang Jajee, Saba and Sannachenkathali was identified as tolerant to water stress with highly accelerated by water stress treatment in the range of 23 to 32% over Control in Catalase (CAT), Super Oxide Dismutase (SOD) and Ascorbate Peroxidase (APX) leads to reduced the cellular membrane damaged by reactive oxygen species and get higher yield; whereas, Matti, Pisang Jajee x Matti, Matti x Anaikomban and Anaikomban x Pisang Jajee were notified as sensitive cultivars and hybrids with lesser increase in antioxidative enzyme activity of 15% than control which is leads to get very low yield.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, ≥20000 units/mg protein, lyophilized powder
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Supelco
Water, suitable for ion chromatography
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, deionized
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, suitable for HPLC