Merck
  • Inicio
  • Resultados de la búsqueda
  • Evaluation of the effect of processing methods on the nutritional and anti-nutritional compositions of two under-utilized Nigerian grain legumes.
Evaluation of the effect of processing methods on the nutritional and anti-nutritional compositions of two under-utilized Nigerian grain legumes.
Pakistan journal of biological sciences : PJBS (2014-02-13)
M O Oke, S S Sobowale, G O Ogunlakin
RESUMEN
The nutritional and anti-nutritional compositions of African Yam Bean (AYB) and Lima bean flours under different processing methods were determined. Nutritional and anti-nutritional properties studied include moisture content, crude protein, crude fibre, ash content, ether extract, carbohydrate, tannin, protease inhibitor and phytate. The moisture content of AYB flours ranged from 9.31 to 9.61% while that of lima beans ranged from 9.32 to 9.56%. There is a significant different among the samples when the unprocessed AYB (control) and the processed AYB were compared. The same trend was also observed with lima bean flours. However, some nutrient did not show significant variations with processing. It was observed that samples of soaked/de-hulled AYB have the least protease inhibitor of 0.73 mg/100 g and it is significantly different from the unprocessed samples. Soaked/de-hulled flours of both AYB and lima beans have the most percentage decrease in anti-nutritional content. Lima bean flours were observed to have higher anti-nutritional content than AYB. The percentage decrease of anti-nutritional factors in the samples is proportionally higher than that of the nutrients. The nutritional and anti-nutritional compositions of the samples suggest that processed African Yam Bean (AYB) and Lima bean flours would have useful application in fabricated foods.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, tested according to Ph. Eur.
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, deionized
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Phytic acid solution, 50 % (w/w) in H2O
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O