• Inicio
  • Resultados de la búsqueda
  • Identification and characterization of uptake systems for cystine and cysteine in cultured astrocytes and neurons: evidence for methylmercury-targeted disruption of astrocyte transport.
Identification and characterization of uptake systems for cystine and cysteine in cultured astrocytes and neurons: evidence for methylmercury-targeted disruption of astrocyte transport.
Journal of neuroscience research (2001-12-18)
G Shanker, M Aschner
RESUMEN
Maintenance of appropriate intracellular glutathione (GSH) levels is crucial for cellular defense against oxidative damage. A suggested mechanism of methylmercury (MeHg) neurotoxicity implicates the involvement of oxygen radical formation and a decrease in cellular levels of GSH. Astrocytes play an important role in providing GSH precursors to neurons, and as will be discussed in this review, altered GSH homeostasis likely leads to impairment of astrocytic handling of glutamate, and neuronal energy metabolism. The review summarizes recent observations on transport systems for cysteine and cystine, precursors of GSH, in primary cultures of astrocytes and neurons, and their sensitivity to MeHg treatment.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
SAFC
L-Cystine
SAFC
L-Cystine
Supelco
L-Cystine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Cystine, certified reference material, TraceCERT®
Cystine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Cystine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.5%