Synthesis of single- and multiple-stranded cystine-rich peptides.
Biopolymers (2004-12-22)
Luis Moroder, Hans-Jürgen Musiol, Marion Götz, Christian Renner
RESUMEN
The large abundance of bioactive single- and multiple-stranded cystine-rich peptides in nature has fostered the development of orthogonal thiol-protection schemes and of efficient chemistries for regioselective disulfide formation in synthetic replica for decades. In parallel to these entirely synthetic strategies, an increased knowledge of oxidative refolding mechanisms of proteins has been accumulated, and the collective experience with air oxidation of cysteine-rich peptides into their native disulfide frameworks have largely confirmed Anfinsen's principle of the self-assembly of polypeptide chains. In fact, a continuously growing number of cysteine-rich bioactive peptides from the most diverse sources and with differing cysteine patterns were found to retain the critical sequence-encoded structural information for correct oxidative folding into the native structures as dominant isomers, although in the biosynthetic pathways the mature peptide forms are mostly generated by posttranslational processing of folded precursors. Such self-assembly processes can be optimized by opportune manipulation of the experimental conditions or by induction of productive intermediates. But there are also numerous cases where folding and disulfide formation are thermodynamically not coupled and where the application of a defined succession of regioselective cysteine pairings still represents the method of choice to install the desired native or non-native cystine frameworks. Among our contributions to the state of the art in the synthesis of cystine-rich peptides, we have mainly addressed the induction of correct oxidative refolding of single-stranded cysteine-rich peptides into their native structures by the use of selenocysteine and suitable strategies for disulfide-mediated assembly of monomers into defined oligomers as mimics of homo- and heterotrimeric collagens as a synthetic approach for the development of new biomaterials.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
SAFC
L-Cystine
SAFC
L-Cystine
Supelco
L-Cystine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Cystine, certified reference material, TraceCERT®
Cystine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Cystine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.5%