Cystine calculi: challenging group of stones.
Postgraduate medical journal (2006-12-07)
Kamran Ahmed, Prokar Dasgupta, Mohammad Shamim Khan
RESUMEN
Cystinuria is an autosomal recessive disorder in renal tubular and intestinal transport of dibasic amino acids, which results in increased urinary excretion of cystine, ornithine, lysine and arginine. It affects 1 in 20 000 people and is caused by a defect in the rBAT gene on chromosome 2. Development of urinary tract cystine calculi is the only clinical manifestation of this disease. Owing to recurrent episodes of stone formation, these patients require a multi-modal approach to management. The role of medical management and minimally invasive surgery was reviewed for the treatment of cystinuria.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
SAFC
L-Cystine
SAFC
L-Cystine
Supelco
L-Cystine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Cystine, certified reference material, TraceCERT®
Cystine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Cystine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.5%