[Dissolution of gallbladder lithiasis with methyl-tert-butyl-ether. Preliminary results: 9 cases].
Presse medicale (Paris, France : 1983) (1988-10-01)
C L Liguory, J F Lefèbvre, D Bonnel, F Cornud
RESUMEN
Nine patients with gallstones were treated by injection of methyl-tert-butyl-ether (MTBE) into the gallbladder. Complete or incomplete dissolution was observed in 5 of the 9 patients. The 4 failures were due to 2 main factors: poor selection of the patients (2 of them had pigment gallstones) and inadequate mixture with the solvent floating above the bile and gallstones. In addition, 4 complications were noted, all of them being resolved by medical treatment. A better selection of the patients and a more efficient technique of MTBE injection should improve these results.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Supelco
tert-Butyl methyl ether, analytical standard
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, 98%
Supelco
tert-Butyl methyl ether, suitable for HPLC, ≥99.8% (GC)