Merck
  • Inicio
  • Resultados de la búsqueda
  • Polymorphisms in sodium-dependent vitamin C transporter genes and plasma, aqueous humor and lens nucleus ascorbate concentrations in an ascorbate depleted setting.
Polymorphisms in sodium-dependent vitamin C transporter genes and plasma, aqueous humor and lens nucleus ascorbate concentrations in an ascorbate depleted setting.
Experimental eye research (2014-05-13)
Srinivasan Senthilkumari, Badri Talwar, Kuppamuthu Dharmalingam, Ravilla D Ravindran, Ramamurthy Jayanthi, Periasamy Sundaresan, Charu Saravanan, Ian S Young, Alan D Dangour, Astrid E Fletcher
RESUMEN
We have previously reported low concentrations of plasma ascorbate and low dietary vitamin C intake in the older Indian population and a strong inverse association of these with cataract. Little is known about ascorbate levels in aqueous humor and lens in populations habitually depleted of ascorbate and no studies in any setting have investigated whether genetic polymorphisms influence ascorbate levels in ocular tissues. Our objectives were to investigate relationships between ascorbate concentrations in plasma, aqueous humor and lens and whether these relationships are influenced by Single Nucleotide Polymorphisms (SNPs) in sodium-dependent vitamin C transporter genes (SLC23A1 and SLC23A2). We enrolled sixty patients (equal numbers of men and women, mean age 63 years) undergoing small incision cataract surgery in southern India. We measured ascorbate concentrations in plasma, aqueous humor and lens nucleus using high performance liquid chromatography. SLC23A1 SNPs (rs4257763, rs6596473) and SLC23A2 SNPs (rs1279683 and rs12479919) were genotyped using a TaqMan assay. Patients were interviewed for lifestyle factors which might influence ascorbate. Plasma vitamin C was normalized by a log10 transformation. Statistical analysis used linear regression with the slope of the within-subject associations estimated using beta (β) coefficients. The ascorbate concentrations (μmol/L) were: plasma ascorbate, median and inter-quartile range (IQR), 15.2 (7.8, 34.5), mean (SD) of aqueous humor ascorbate, 1074 (545) and lens nucleus ascorbate, 0.42 (0.16) (μmol/g lens nucleus wet weight). Minimum allele frequencies were: rs1279683 (0.28), rs12479919 (0.30), rs659647 (0.48). Decreasing concentrations of ocular ascorbate from the common to the rare genotype were observed for rs6596473 and rs12479919. The per allele difference in aqueous humor ascorbate for rs6596473 was -217 μmol/L, p < 0.04 and a per allele difference in lens nucleus ascorbate of -0.085 μmol/g, p < 0.02 for rs12479919. The β coefficients for the regression of log10 plasma ascorbate on aqueous humor ascorbate were higher for the GG genotype of rs6596473: GG, β = 1460 compared to carriage of the C allele, CG, β = 1059, CC, β = 1132, p interaction = 0.1. In conclusion we found that compared to studies in well-nourished populations, ascorbate concentrations in the plasma, aqueous humor and lens nucleus were low. We present novel findings that polymorphisms in SLC23A1/2 genes influenced ascorbate concentration in aqueous humor and lens nucleus.
MATERIALES
Referencia del producto
Marca
Descripción del producto
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Sodium ascorbate, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
(+)-Sodium L-ascorbate, analytical standard
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, 99%
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®