Kinetics and utilization of lipid sources during acute exercise and acipimox.
American journal of physiology. Endocrinology and metabolism (2014-06-05)
Birgitte Nellemann, Esben Søndergaard, Jørgen Jensen, Steen Bønløkke Pedersen, Niels Jessen, Jens Otto Lunde Jørgensen, Søren Nielsen
RESUMEN
Overweight is associated with abnormalities of lipid metabolism, many of which are reversed by exercise. We investigated the impact of experimental antilipolysis and acute exercise on lipid kinetics and oxidation from VLDL-TG, plasma FFA, and "residual lipids" in overweight men (n = 8) using VLDL-TG and palmitate tracers in combination with muscle biopsies in a randomized, placebo-controlled design. Participants received placebo or acipimox on each study day (4 h of rest, 90 min of exercise at 50% V(O(2 max))). Exercise suppressed VLDL-TG secretion significantly during placebo but not acipimox (placebo-rest: 64.2 ± 9.4; placebo-exercise: 48.3 ± 8.0; acipimox-rest: 55.2 ± 13.4; acipimox-exercise: 52.0 ± 10.9). Resting oxidation of VLDL-TG FA and FFA was significantly reduced during acipimox compared with placebo, whereas "residual lipid oxidation" increased significantly [VLDL-TG oxidation (placebo: 18 ± 3 kcal/h; acipimox: 11 ± 2 kcal/h), FFA oxidation (placebo: 14 ± 2 kcal/h; acipimox: 4 ± 0.5 kcal/h), and residual lipid oxidation (placebo: 3 ± 5 kcal/h; acipimox: 14 ± 5 kcal/h)]. Additionally, during exercise on both placebo and acipimox, oxidation of VLDL-TG and FFA increased, but the relative contribution to total lipid oxidation diminished, except for FFA, which remained unchanged during acipimox. Residual lipid oxidation increased significantly during exercise in both absolute and relative terms. Changes in selected cellular enzymes and proteins provided no explanations for kinetic changes. In conclusion, suppressed FFA availability blunts the effect of exercise on VLDL-TG secretion and modifies the contribution of lipid sources for oxidation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
HEPES
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Supelco
Palmitic acid, analytical standard
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
HEPES
Sigma-Aldrich
Serum Triglyceride Determination Kit,  kit sufficient for 250 tests
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E422, anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Palmitic acid, ≥95%, FCC, FG
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)