Merck
Cholesterol metabolism and immunity.
The New England journal of medicine (2014-11-13)
Anna Simon
RESUMEN
A study in mice implicates a cholesterol derivative in damping down the inflammatory response mediated by interleukin-1β and explaining, at least in part, the immunosuppressive effect of type I interferon, which is used in the treatment of multiple sclerosis.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
25-Hydroxycholesterol, ≥98%