Merck
Salmeterol's extreme β2 selectivity is due to residues in both extracellular loops and transmembrane domains.
Molecular pharmacology (2014-10-18)
Jillian G Baker, Richard G W Proudman, Stephen J Hill
RESUMEN
Salmeterol is a long-acting β2-agonist, widely used as an inhaled treatment of asthma and chronic obstructive pulmonary disease. It has very high β2-affinity (log KD -8.95) and is very selective for the β2-adrenoceptor (1000-fold selectivity over the β1-adrenoceptor). This study used a mutagenesis approach to determine the exact amino acids in the human β2-adrenoceptor responsible for this very high selectivity. Wild-type β2- and β1-adrenoceptors, chimeric β2/β1-adrenoceptors, and receptors with single-point mutations were transfected into Chinese hamster ovary-K1 cells, and affinity and function were studied using [(3)H]CGP 12177 [(-)-4-(3-tert-butylamino-2-hydroxypropoxy)-benzimidazol-2-one] whole-cell binding and [(3)H]cAMP accumulation. Extracellular loop 3 (and specifically amino acid K305) had the largest single effect by reducing salmeterol's affinity for the β2-adrenoceptor by 31-fold. H296 in transmembrane 6 also had a major effect (18-fold reduction in salmeterol affinity). Combining these, in the double mutant β2-H296K-K305D, reduced salmeterol's affinity by 275-fold, to within 4-fold of that of the β1-adrenoceptor, without affecting the affinity or selectivity of other β2-agonists (salbutamol, formoterol, fenoterol, clenbuterol, or adrenaline). Another important amino acid was Y308 in transmembrane 7, although this also affected the affinity and selectivity of other agonists. F194 in extracellular loop 2 and R304 in extracellular loop 3 also had minor effects. None of these mutations (including the double mutant β2-H296K-K305D) affected the efficacy or duration of action of salmeterol. This suggests that the high affinity and selectivity of salmeterol are due to specific amino acids within the receptor itself, but that the duration of action is at least in part due to other factors, for example lipophilicity.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Supelco
Albuterol sulfate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Adenine, Pharmaceutical Secondary Standard; Certified Reference Material
Adenine, European Pharmacopoeia (EP) Reference Standard
Salbutamol sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Supelco
Salbutamol, VETRANAL®, analytical standard
Sigma-Aldrich
Salbutamol hemisulfate salt, ≥98%
Supelco
Salbutamol hemisulfate salt, VETRANAL®, analytical standard
Supelco
Aluminum oxide, for the determination of hydrocarbons
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Salbutamol
Sigma-Aldrich
Aluminum oxide, pellets, 3 mm
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, calcined, powder, primarily α-phase, 100-325 mesh
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, activated, basic, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I