Access to large cyclic peptides by a one-pot two-peptide segment ligation/cyclization process.
Organic letters (2014-12-17)
Emmanuelle Boll, Jean-Philippe Ebran, Hervé Drobecq, Ouafâa El-Mahdi, Laurent Raibaut, Nathalie Ollivier, Oleg Melnyk
RESUMEN
The use of the N-acetoacetyl protecting group for N-terminal cysteine residue enabled creation of an efficient and mild one-pot native chemical ligation/SEA ligation sequence giving access to large cyclic peptides.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
SAFC
L-Cysteine
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Supelco
L-Cysteine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%