Human testicular peritubular cells host putative stem Leydig cells with steroidogenic capacity.
The Journal of clinical endocrinology and metabolism (2014-04-02)
Luise Landreh, Katrin Spinnler, Kerstin Schubert, Merja R Häkkinen, Seppo Auriola, Matti Poutanen, Olle Söder, Konstantin Svechnikov, Artur Mayerhofer
RESUMEN
We aim to examine the steroidogenic phenotype and the differentiation potential of human testicular peritubular cells (HTPCs) and to explore their possible relationship to the adult Leydig cell lineage. The cells of the adult Leydig cell lineage may reside in the peritubular compartment of the testis. This suggestion is supported by the facts that the rodent peritubular cells can be differentiated toward this lineage and that cAMP enhances their steroidogenic potential. Human testicular biopsies, and derived HTPCs, were analyzed by immunohistochemistry, RT-PCR, and Western blotting. After stimulation by forskolin or platelet-derived growth factor-BB, quantitative RT-PCR was used to compare the levels of mRNAs encoding proteins involved in steroidogenesis and steroid production was analyzed by liquid chromatography and tandem mass spectrometry. Immunohistochemical analysis revealed that the peritubular cells that form the outer part of the tubular wall express platelet derived growth factor receptor-α. Furthermore, the pluripotency markers (POU domain class 5 transcription factor 1, GATA-binding protein 4), stem Leydig cell markers (platelet derived growth factor receptor-A, leukemia inhibitory factor receptor), and mRNAs encoding proteins involved in steroidogenesis (nuclear receptor subfamily 5, group A, member 1; steroidogenic acute regulatory protein; CYP11A1; CYP17A1; 3β-hydroxysteroid dehydrogenase) were expressed by the HTPCs. Stimulation with forskolin increased the expression of the steroidogenic markers, which was accompanied by the production of pregnenolone and progesterone by HTPCs in vitro. Treatment with platelet-derived growth factor-BB induced expression of steroidogenic acute regulatory protein. Our results indicate that the tubular wall of the human testis is a reservoir for cells of the adult Leydig cell lineage and that the steroidogenic potential of these cells can be activated in culture.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
SAFC
HEPES
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
PDGF-BB human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Progesterone, European Pharmacopoeia (EP) Reference Standard
Redes sociales
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Investigación. Desarrollo. Producción.
Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos
© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.
Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.