Long non coding RNA-UCA1 contributes to cardiomyocyte apoptosis by suppression of p27 expression.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-04-15)
Youbin Liu, Daliang Zhou, Guangnan Li, Xing Ming, Ying feng Tu, Jinwei Tian, Huimin Lu, Bo Yu
RESUMEN
Urothelial carcinoma-associated 1 (UCA1) is a recently identified long non coding RNA (lncRNA). However, few studies have explored its role in cardiomyocytes after focal cardiac ischemia reperfusion injury (CIR). Rat CIR models were established using ligation of the Lower Anterior Descending artery (LAD). Cell apoptosis and reactive oxygen species (ROS) production in cardiac tissues were explored using immunohistochemistry and DHE staining. lncRNA expression patterns were detected using microarray and validated by qPCR. Cell viability and apoptosis were examined using MTT assay and flow cytometry. CIR significantly induced cell apoptosis and ROS production in the rat model. The results of microarray demonstrated the reduced expression of UCA1, which was validated by qPCR. Follow-up experiments showed that UCA1 was involved in H2O2-induced cell apoptosis. We further showed that UCA1 negatively correlated with the expression of p27. Moreover, overexpression of p27 could induce primary cardiomyocyte apoptosis. Reduction of UCA1 levels plays a pro-apoptotic role in primary cardiomyocytes partially through stimulation of p27 protein expression. These results are in agreement with the observed levels of UCA1, p27 and apoptosis after cardiac I/R injury, suggesting that UCA1 might have an important role during I/R injury.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
2,3,5-Triphenyltetrazolium chloride, ≥98.0% (HPLC)
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Protease Inhibitor Cocktail, Animal Component Free, for use with mammalian cell and tissue extract, DMSO solution
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)