• Inicio
  • Resultados de la búsqueda
  • The study of binding of methyl tert-butyl ether to human telomeric G-quadruplex and calf thymus DNA by gas chromatography, a thermodynamic discussion.
The study of binding of methyl tert-butyl ether to human telomeric G-quadruplex and calf thymus DNA by gas chromatography, a thermodynamic discussion.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-10-04)
Sahar Ghasemi, Farhad Ahmadi
RESUMEN
Methyl-tert-butyl ether (MTBE) is widely used as an antiknock additive for increasing octane number of gasoline. Recently, the in vivo studies demonstrated that MTBE has genotoxic potential and able to form adducts with DNA. In the work, the interactions of MTBE with calf thymus DNA (ct-DNA) and the Na(+) form of G-quadruplex DNA (wtTel22) were studied by using of head space-solid phase microextraction technique coupled to gas chromatography. The binding equilibrium constants were measured through the equilibriums of a four phase system. In addition, the MTBE Henry's law constants for two different buffers in the temperature range of 283-303K were measured. Thermodynamic studies revealed that the complexation of MTBE to both DNAs is enthalpy favored and entropy disfavored. The thermodynamic results revealed that MTBE may have interaction with ct-DNA via the minor groove of DNA. Also, MTBE may be complexed into the basket of G-quadruplex structure. In addition, the low difference in the binding constants of MTBE for both different DNA targets may confirm that MTBE is poorly selective for different conformations of DNA.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
Dibutyl ether, anhydrous, 99.3%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
Supelco
tert-Butyl methyl ether, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Dibutyl ether, ReagentPlus®, ≥99%
Supelco
tert-Butyl Methyl Ether, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME/HPLC Interface, rheodyne valve version
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, 98%
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
tert-Butyl methyl ether, suitable for HPLC, ≥99.8% (GC)