• Inicio
  • Resultados de la búsqueda
  • A novel method for the simultaneous analysis of seven biothiols in rice (Oryza sativa L.) using hydrophilic interaction chromatography coupled with electrospray tandem mass spectrometry.
A novel method for the simultaneous analysis of seven biothiols in rice (Oryza sativa L.) using hydrophilic interaction chromatography coupled with electrospray tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-02)
Zhao-Yun Cao, Li-Hua Sun, Ren-Xiang Mou, Rong Zhou, Zhi-Wei Zhu, Ming-Xue Chen
RESUMEN
Analysis of biothiols is still problematic, due to their high polarity, oxidation sensitivity and time-consuming sample preparation. In this paper, a direct, rapid and sensitive method was developed for simultaneous quantification of unbound cysteine (Cys), glutathione (GSH) and phytochelatins (PCs) in rice leaf, stem and root samples by hydrophilic interaction chromatography coupled with electrospray tandem mass spectrometry (HILIC-MS/MS). Homogenized samples were extracted with water containing 50mM dithiothreitol, without derivatization and further clean-up, and the extracts were injected directly onto an Xbridge Amide-HILIC column (3.5μm, 150mm×2.1mm i.d.). The best chromatographic separation and MS sensitivity was achieved using a linear gradient elution with 10mM aqueous ammonium formate and acetonitrile as the mobile phase. In MS/MS mode the detection limit (S/N≥3) of seven biothiols was 3-105nM. Good linearities were observed (r>0.995) with linear dynamic range at least over three orders of magnitude. Recoveries for most analytes were within the range of 77-128%, with relative standard deviations less than 18.2%. The intra-day precision (n=7) was 6.1-11.7%, and the inter-day precision over 15 d (n=15) was 8.5-16.3% for all biothiols. The optimized HILIC-MS/MS method was applied to study the influence of different cadmium (Cd) concentrations (0, 1 and 50μM) on contents of Cys, GSH and PC2-6 in rice tissue. With increasing Cd concentrations in nutrient solutions, contents of PC2-4 in rice roots increased but contents of Cys and GSH decreased. Contents of PC2-4 in both rice leafs and stems increased markedly at high dose Cd (50μM) treatment compared with controls, compared with low Cd concentrations (1μM). However, both PC5 and PC6 were not detected throughout the stress experiment.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
L-Glutathione reduced, BioReagent, suitable for cell culture, ≥98.0%, powder
SAFC
L-Cysteine
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Supelco
L-Cysteine, certified reference material, TraceCERT®
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis