Potential analytical interferences and seasonal variability in diethyltoluamide environmental monitoring programs.
Chemosphere (2015-03-10)
Sylvain Merel, Andrey I Nikiforov, Shane A Snyder
RESUMEN
N,N-diethyl-m-toluamide (DEET), the active component of many insect repellents, is among the most frequently detected compounds in aqueous environments with concentrations reported in the ng L(-1) to μg L(-1) range. However, DEET is often detected in blanks and reported concentrations differ significantly depending on the analytical technique employed. In addition, relatively sparse data are available regarding the seasonal variability of DEET concentrations in water and there are apparent inconsistencies with expected use patterns. Therefore, the present study investigates potential interferences affecting the detection and quantification of DEET then the geographical and seasonal variations of DEET concentrations. To examine potential analytical interferences, DEET was analyzed in five geographically-diverse wastewater effluents using both gas chromatography and liquid chromatography coupled to mass spectrometric detectors. At times, the concentrations quantified by the employed analytical methods varied significantly. Five compounds with similar molecular weights and structures as DEET were investigated as potential mimics and some were shown to induce an overestimation of DEET. Further experimentation suggested that the solvents used in sample preparation and HPLC analysis are another possible source of interference. Besides potential interferences, the seven-month weekly monitoring of DEET in the primary effluent of a wastewater treatment plant demonstrated a clear seasonal trend with decreasing concentration from summer to winter. These data collectively demonstrate that there are many challenges in the quantification of DEET in complex environmental samples and that co-occurrence of similarly structured substances present in the water sample and/or the solvents used for the analysis could induce analytical bias.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)