Merck
  • Inicio
  • Resultados de la búsqueda
  • Evaluation of the structural composition and surface properties of rhamnolipid mixtures produced by Pseudomonas aeruginosa UFPEDA 614 in different cultivation periods.
Evaluation of the structural composition and surface properties of rhamnolipid mixtures produced by Pseudomonas aeruginosa UFPEDA 614 in different cultivation periods.
Applied biochemistry and biotechnology (2014-10-30)
Arquimedes Paixão de Santana-Filho, Doumit Camilios-Neto, Lauro Mera de Souza, Guilherme Lanzi Sassaki, David Alexander Mitchell, Nadia Krieger
RESUMEN
We studied the production of rhamnolipids by Pseudomonas aeruginosa UFPEDA 614 in submerged culture, using glycerol as the carbon source. A rhamnolipid yield of 15.9 g/L was obtained with 40 g/L glycerol and 5 g/L sodium nitrate as nitrogen source after 7 days of cultivation. Structural analysis carried out at different cultivation periods showed that the four major mono-rhamnolipid homologues are present in higher proportion in the first 48 h. Over time, the corresponding four major di-rhamnolipid homologues predominated, representing about 75 % of the total rhamnolipids after 96 h. Physicochemical analysis of the rhamnolipid mixtures obtained at different cultivation periods showed that the sample obtained from the first day of cultivation had the lower critical micelle concentration (15.6 mg/L), which is probably related to the higher proportion of mono-rhamnolipids. The results presented here show that the composition of the mixture of rhamnolipid homologues produced by P. aeruginosa UFPEDA 614 varies over time and that this variation influences the physicochemical properties of the mixture. These findings can be used in order to produce rhamnolipid mixtures that have suitable properties for different applications.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Sigma-Aldrich
Glycerol, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E422, anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Ammonium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N