Merck
ZnO-modified cellulose fiber sheets for antibody immobilization.
Carbohydrate polymers (2014-05-13)
Vinay Khatri, Katalin Halász, Lidija V Trandafilović, Suzana Dimitrijević-Branković, Paritosh Mohanty, Vladimir Djoković, Levente Csóka
RESUMEN
Cellulose fiber sheets impregnated with saccharide capped-ZnO nanoparticles were used as bioactive materials for antibody immobilization. First, ZnO nanoparticles were synthesized in the presence of glucose (monosaccharide), sucrose (disaccharide) as well as alginic acid and starch (polysaccharides). The pine cellulose fibers were then modified by the obtained saccharide capped nanoparticles and further incorporated into the sheets. The presence of ZnO significantly improved the immobilization of the antibodies on the surface of the sheets. After rewetting the alginic acid-ZnO modified sheets with saline solution, the retention of antibodies was about 95%. A high degree of the immobilization of biomolecules is an important feature for possible fabrications of bioactive- or biosensing-papers and we successfully tested the sheets on the detection of blood types using (A, B, and D blood antibodies). The ZnO nanoparticles affected also the other properties of the sheets. The ZnO-modified fiber sheets showed higher values of tensile index (strength), smoothness and opacity, while the value of porosity was substantially lower than that of the unmodified sheet. The presence of ZnO nanoparticles provided also the antimicrobial activity to the sheets. They showed a strong activity against bacteria (Escherichia coli and Staphylococcus aureus) and strong resistance to the attack of cellulase producing fungus Gloeophyllum trabeum.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 20-40 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)