Merck
  • Inicio
  • Resultados de la búsqueda
  • Are cyclic lipopeptides produced by Bacillus amyloliquefaciens S13-3 responsible for the plant defence response in strawberry against Colletotrichum gloeosporioides?
Are cyclic lipopeptides produced by Bacillus amyloliquefaciens S13-3 responsible for the plant defence response in strawberry against Colletotrichum gloeosporioides?
Letters in applied microbiology (2014-12-17)
S Yamamoto, S Shiraishi, S Suzuki
RESUMEN
The antagonistic strain Bacillus amyloliquefaciens strain S13-3 decreased the severity of strawberry anthracnose caused by Colletotrichum gloeosporioides. The foliar application of S13-3 triggered the expression of pathogenesis-related proteins, chitinase and β-1,3-glucanase, in strawberry leaves. We identified lipopeptide antibiotics, including iturin A, fengycin, mixirin, pumilacidin and surfactin, produced and secreted by S13-3. Iturin A and surfactin elicited the gene expression of the pathogenesis-related proteins in strawberry leaves, suggesting that antagonistic strain S13-3 confers resistance to strawberry leaves through the production of lipopeptide antibiotics. In fact, iturin A and surfactin triggered induced systemic resistance on strawberry plants, resulting in the reduction of the severity of anthracnose disease caused by Colletotrichum gloeosporioides. The bifunctional activity of S13-3, which consists of the antagonistic effect and the induction of plant defence response by the antibiotics produced by it, may make S13-3 an innovative biological control agent against phytopathogens in strawberry. This study tries to determine whether biocontrol of phytopathogens by Bacillus amyloliquefaciens in strawberry can be connected to induced plant resistance. The results suggested that the antagonistic strain B. amyloliquefaciens S13-3 confers resistance to strawberry through the production of lipopeptide antibiotics.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Supelco
Sodium acetate solution, NMR reference standard, 50 mM in D2O (99.9 atom % D), water 1 %, NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Supelco
Acetonitrile, analytical standard
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%