• Inicio
  • Resultados de la búsqueda
  • Liposomes modified with superhydrophilic polymer linked to a nonphospholipid anchor exhibit reduced complement activation and enhanced circulation.
Liposomes modified with superhydrophilic polymer linked to a nonphospholipid anchor exhibit reduced complement activation and enhanced circulation.
Journal of pharmaceutical sciences (2014-11-14)
Okhil K Nag, Vivek R Yadav, Brandon Croft, Andria Hedrick, Vibhudutta Awasthi
RESUMEN
We report the synthesis of an acyl-anchored superhydrophilic polymer (SHP) for external surface modification of liposome surface. N¹-(2-aminoethyl)-N⁴-hexadecyl-2-tetradecylsuccinamide conjugated with SHP (HDAS-SHP) was synthesized and used for modifying the liposome surface. Unlike polyethylene glycol (PEG)-phospholipids, which are commonly used for manufacturing stealth liposomes, HDAS-SHP is devoid of both PEG and phosphoryl groups and possesses a zwitterionic polymeric chain. Circulation persistence of the ⁹⁹(m)Tc-labeled HDAS-SHP liposomes was documented by gamma camera imaging. After 24 h postinjection, approximately 30% of injected HDAS-SHP liposomes were present in blood as compared with only 4.5% of the plain liposomes. HDAS-SHP liposomes inhibited complement activation. They were found to be amenable to pH-gradient-based active loading of Adriamycin in a stable manner. At 37°C, HDAS-SHP liposomes provided better encapsulation efficiencies than the liposomes modified with DSPE-PEG₂₀₀₀. These results provide a strong basis for HDAS-SHP as a viable alternative to PEG-phospholipids for imparting stealth characteristics to drug delivery vehicles such as liposomes. © 2014 Wiley Periodicals, Inc. and the American Pharmacists Association J Pharm Sci 104:114-123, 2015.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
SAFC
Cholesterol, Plant-Derived
Supelco
Dichloromethane, analytical standard