Merck
Regulation of ubiquitin-specific processing protease 8 suppresses neuroinflammation.
Molecular and cellular neurosciences (2014-05-28)
Lihong Zhu, Wei Bi, Dan Lu, Chanjuan Zhang, Xiaoming Shu, Huadong Wang, Renbing Qi, Qiaoyun Shi, Daxiang Lu
RESUMEN
In our previous study, we reported that luteolin might exert neuroprotective functions by inhibiting the production of proinflammatory mediators, thereby suppressing microglial activation. In this study, we used two-dimensional gel electrophoresis (2-DE) and mass spectrometry (MS) to study the effect of ubiquitin-specific processing protease 8 (USP8) in luteolin-treated microglia. Western blot analysis verified that USP8 expression is upregulated by luteolin. Researchers have found that USP8 markedly enhanced the stability of neuregulin receptor degradation protein-1 (Nrdp1), which in turn inhibited the production of proinflammatory cytokines in toll-like receptor-triggered macrophages. We next hypothesized that luteolin inhibits microglial inflammation by regulating USP8 gene expression. After transfecting BV2-immortalized murine microglial cells with USP8, a significant reduction in the degradation of Nrdp1 was observed. USP8 overexpression also reduced the production of lipopolysaccharide (LPS)-induced proinflammatory mediators such as inducible nitric oxide synthase (iNOS), nitric oxide (NO), cyclooxygenase-2 (COX-2), and prostaglandin E2 (PGE2). We also found that USP8 siRNA blocked luteolin inhibition of pro-inflammatory gene expression such as iNOS, NO, COX-2, and PGE2. Taken together, our findings suggested that luteolin inhibits microglial inflammation by enhancing USP8 protein production. We concluded that in addition to anti-inflammatory luteolin, USP8 might represent a novel mechanism for the treatment of neuroinflammation and neurodegeneration.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, ACS reagent
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent