Merck
Interferon-tau attenuates uptake of nanoparticles and secretion of interleukin-1β in macrophages.
PloS one (2014-12-09)
Kyoko Hara, Koumei Shirasuna, Fumitake Usui, Tadayoshi Karasawa, Yoshiko Mizushina, Hiroaki Kimura, Akira Kawashima, Akihide Ohkuchi, Shuichi Matsuyama, Koji Kimura, Masafumi Takahashi
RESUMEN
Type I interferons (IFNs), including IFN-alpha (IFNA) and IFN-beta (IFNB), have anti-inflammatory properties and are used to treat patients with autoimmune and inflammatory disorders. However, little is known of the role of IFN-tau (IFNT), a type I IFN produced by ruminant animals for inflammation. Because IFNB has recently been shown to inhibit nucleotide-binding oligomerization domain-like receptor, pyrin domain-containing 3 (NLRP3) inflammasome activation and subsequent secretion of the potent inflammatory cytokine interleukin (IL)-1β, we examined the effects of ruminant IFNT on NLRP3 inflammasome-mediated IL-1β secretion in human THP-1 macrophages. IFNT dose-dependently inhibited IL-1β secretion induced by nano-silica, a well-known activators of NLRP3 inflammasomes, in human macrophages primed with lipopolysaccharide (LPS, TLR4 agonist) and Pam3CSK4 (TLR1/2 agonist). IFNT also suppressed phagocytosis of nano-silica and reactive oxygen species (ROS) generation. Western blot analysis showed that IFNT inhibited both pro-IL-1β and mature IL-1β. In addition, real-time RT-PCR analysis showed that IFNT suppressed IL-1β mRNA expression induced by LPS and Pam3CSK4. Although nano-silica particles did not induce IL-10 secretion, IFNT induced IL-10 secretion in a dose-dependent manner. Furthermore, IFNT-suppressed IL-1β secretion was restored by anti-IL-10 neutralizing antibody. Ruminant IFNT inhibits NLRP3 inflammasome-driven IL-1β secretion in human macrophages via multiple pathways, including the uptake of nano-silica particles, generation of ROS, and IL-10-mediated inhibition of pro-IL-1β induction. It may be a therapeutic alternative to IFNA and IFNB.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Adenosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis