Merck
miR-20a inhibits TCR-mediated signaling and cytokine production in human naïve CD4+ T cells.
PloS one (2015-04-18)
Amarendra V Reddycherla, Ines Meinert, Annegret Reinhold, Dirk Reinhold, Burkhart Schraven, Luca Simeoni
RESUMEN
Upon TCR stimulation by peptide-MHC complexes, CD4+ T cells undergo activation and proliferation. This process will ultimately culminate in T-cell differentiation and the acquisition of effector functions. The production of specific cytokines by differentiated CD4+ T cells is crucial for the generation of the appropriate immune response. Altered CD4+ T-cell activation and cytokine production result in chronic inflammatory conditions and autoimmune disorders. miRNAs have been shown to be important regulators of T-cell biology. In this study, we have focused our investigation on miR-20a, a member of the miR-17-92 cluster, whose expression is decreased in patients suffering from multiple sclerosis. We have found that miR-20a is rapidly induced upon TCR-triggering in primary human naïve CD4+ T cells and that its transcription is regulated in a Erk-, NF-κB-, and Ca++-dependent manner. We have further shown that overexpression of miR-20a inhibits TCR-mediated signaling but not the proliferation of primary human naïve CD4+ T cells. However, miR-20a overexpression strongly suppresses IL-10 secretion and moderately decreases IL-2, IL-6 and IL8 production, which are crucial regulators of inflammatory responses. Our study suggests that miR-20a is a new player in the regulation of TCR signaling strength and cytokine production.
MATERIALES
Referencia del producto
Marca
Descripción del producto
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%