Merck
Vitrified collagen-based conjunctival equivalent for ocular surface reconstruction.
Biomaterials (2014-06-17)
Huifang Zhou, Qiaozhi Lu, Qiongyu Guo, Jemin Chae, Xianqun Fan, Jennifer H Elisseeff, Michael P Grant
RESUMEN
The main functions of the conjunctiva, an essential part of the ocular surface, are to maintain the equilibrium of the tear film and to protect the eye. Upon injuries, the prerequisite to successful ocular surface repair is conjunctival reconstruction. Tissue engineering techniques, including transplantation of autografts, amniotic membranes and numerous synthetic/natural materials, have been developed. However, none of these strategies is completely satisfactory due to lack of goblet cell repopulation, poor mechanical properties or non-standardized preparation procedure. Here, we cultured conjunctival epithelial cells on vitrified collagen membranes and developed a tissue equivalent for repairing damaged conjunctiva. Optimized vitrified collagen has superior mechanical and optical properties to previous biomaterials for ocular surface application, and its unique fibrillar structure significantly benefited conjunctival epithelial cell growth and the phenotypic development in vitro. In a rabbit model, vitrified collagen greatly promoted conjunctival regeneration with rapid re-epithelization, sufficient repopulation of goblet cells and minimized fibrosis and wound contracture, proved by gene expression analyses and histological staining. In conclusion, we have demonstrated the potential suitability of utilizing vitrified collagen-based tissue equivalent in ocular surface reconstruction.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC), suitable for fluorescence
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Diethyl azodicarboxylate solution, 40 wt. % in toluene
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
SAFC
HEPES
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
HEPES
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O