Merck
  • Inicio
  • Resultados de la búsqueda
  • A validated liquid chromatography-tandem mass spectrometry method for the determination of methyl gallate and pentagalloyl glucopyranose: application to pharmacokinetic studies.
A validated liquid chromatography-tandem mass spectrometry method for the determination of methyl gallate and pentagalloyl glucopyranose: application to pharmacokinetic studies.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-02-24)
Pimsumon Jiamboonsri, Pimolpan Pithayanukul, Rapepol Bavovada, Song Gao, Ming Hu
RESUMEN
Methyl gallate (MG) and pentagalloyl glucopyranose (PGG) are bioactive phenolic compounds that are widely distributed in herbs and plant foods. Their potential activities include anti-oxidant, anti-inflammatory, anti-cancer, anti-bacterial and anti-viral activities. However, knowledge concerning the pharmacokinetic characteristics of MG and PGG is limited. The purpose of this study was to develop a sensitive and reproducible ultra-performance liquid chromatography-tandem mass spectrometric (UPLC-MS/MS) method to simultaneously quantify MG and PGG in rat blood samples. The linear response ranges for MG and PGG were 0.0195-20 and 0.0390-20 μM, respectively. The lower limit of quantification was 0.0195 μM for MG and 0.0390 μM for PGG. The intra- and inter-day variances were less than 15%, and accuracy was within 80-120%. This assay was successfully applied to pharmacokinetic studies in Sprague-Dawley rats after intraperitoneal administration of MG and PGG (20 mg/kg). The values of areas under the blood concentration time curves (AUC₀₋₂₄ h) for MG and PGG were 109.9 ± 73.40 and 38.78 ± 24.53 h*μM, respectively. The maximum blood concentrations (Cmax) of MG and PGG were 34.72 ± 17.32 and 6.39 ± 4.25 μM, respectively. The time required to reach the maximum concentration (Tmax) was 0.85 ± 0.70 h for both MG and PGG. The values of the elimination rate constant (Ke), elimination half-life (t1/2), volume of distribution (Vd), clearance (Cl) and mean resident time (MRTlast) were 0.056 ± 0.032 h(-1), 17.50 ± 12.25 h, 530.95 ± 247.54 L/kg, 159.91±76.05L/h/kg, 8.71 ± 2.53 h for MG and 0.023 ± 0.012 h(-1), 38.66 ± 22.89 h, 7838.89 ± 3474.72 L/kg, 30.98 ± 21.73 L/h/kg, 12.47 ± 2.77 h for PGG, respectively. In conclusion, a UPLC-MS/MS method was fully validated over a wide linear range and used to quantify the levels of MG and PGG in pharmacokinetic studies of MG and PGG in rats. The main advantages of this method are the use of small blood volumes (10 μL), rapid analysis (5 min) and excellent recoveries.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Penta-O-galloyl-β-D-glucose hydrate, ≥96% (HPLC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methyl 3,4,5-trihydroxybenzoate, 98%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles