Merck
Evaluation of a compact mass spectrometer for routine support of pharmaceutical chemistry.
Journal of pharmaceutical and biomedical analysis (2014-03-04)
Xiaodong Bu, Jiong Yang, Xiaoyi Gong, Christopher J Welch
RESUMEN
The suitability of a recently introduced inexpensive, compact mass spectrometer detector is evaluated for supporting pharmaceutical chemistry investigations. While high performance/high cost MS detectors dominate the marketplace, there is growing recognition of the need for a small, inexpensive MS detector with reduced capabilities for supporting synthetic chemistry investigations, where reduced sensitivity and unit mass resolution are often suitable for solving routine problems. In this study, the fundamental performance characteristics of the recently introduced Advion compact mass spectrometer were evaluated, investigating the use of the instrument for routine product and impurity identification, reaction monitoring, evaluation of potential genotoxic impurities and study of high molecular weight biomolecules. In general, the results of the evaluation show this compact and inexpensive mass spectrometer to be well suited for providing reliable support for pharmaceutical chemistry investigations, with sub-nanogram limit of detection and impurity identification below 0.1% being possible in some instance.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%