• Inicio
  • Resultados de la búsqueda
  • Comparative evaluation of seven different sample treatment approaches for large-scale multiclass sport drug testing in urine by liquid chromatography-mass spectrometry.
Comparative evaluation of seven different sample treatment approaches for large-scale multiclass sport drug testing in urine by liquid chromatography-mass spectrometry.
Journal of chromatography. A (2014-08-21)
Juan C Domínguez-Romero, Juan F García-Reyes, Antonio Molina-Díaz
RESUMEN
Sample preparation is a critical step in large-scale multiclass analysis such as sport drug testing. Due to the wide heterogeneity of the analytes and the complexity of the matrix, the selection of a correct sample preparation method is essential, looking for a compromise between good recoveries for most of the analytes and cleanliness of the extract. In the present work, seven sample preparation procedures based on solid-phase extraction (SPE) (with 5 different cartridges), liquid-liquid extraction (LLE) and sorbent-supported liquid extraction (SLE) were evaluated for multiclass sport drug testing in urine. The selected SPE sorbents were polymeric cartridges Agilent PLEXA™ and Oasis HLB™, mixed mode cation and anion exchange cartridges Oasis MAX™ and MCX™, and C18 cartridges. LLE was performed using tert-butyl methyl ether and SLE was carried out using Agilent Chem Elut™ cartridges. To evaluate the proposed extraction procedures, a list of 189 compounds were selected as representative from different groups of doping agents, including 34 steroids, 14 glucocorticosteroids, 24 diuretics and masking agents, 11 stimulants, 9 beta-agonist, 16 beta-blockers, 6 Selective Estrogen Receptors Modulators (SERMs), 24 narcotics and 22 other drugs of abuse/sport drugs. Blank urine samples were spiked at two levels of concentration, 2.5 and 25μgL(-1) and extracted with the different extraction protocols (n=6). The analysis of the extracts was carried out by liquid chromatography electrospray time-of-flight mass spectrometry. The use of solid-phase extraction with polymer cartridges provided high recoveries for most of the analytes tested and was found the more suitable method for this type of application given the additional advantages such as low sample and solvent consumption along with increased automation and throughput.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%