Investigation of the influence of vanadium compounds treatment in NZO mice model--preliminary study.
Acta poloniae pharmaceutica (2014-10-03)
Mirosław Krośniak, Renata Francik, Katarzyna Kołodziejczyk, Agnieszka Wojtanowska-Krośniak, Cinzia Tedeschi, Veronica Petrone, Ryszard Gryboś
RESUMEN
Abstract: New Zealand obese mice (NZO) are characterized by symptoms similar to human metabolic syndrome. Vanadium in different investigations showed anti-diabetic activity but until now an NZO mice model has not been tested with this element. The aim of this study was to investigate anti-diabetic activity of three vanadium compounds (VOSO4, VO(mal)2 and Na(VO(O2)2bpy) x 8H2O) in the NZO model. Metabolic syndrome was induced by special diet (1.5% of cholesterol and 15% of saturated fatty acids) during 8 weeks. In the next 5 weeks, the tested vanadium compounds were administered once daily, in a dose of 0.063 mmol/kg of body mass. At the end of the experiment, glucose, cholesterol, triglycerides and alanine transaminase were measured in the serum. The obtained results showed that the glucose level was decreased nearly to the healthy NZO mice in comparison to the NZO mice with metabolic syndrome. In all groups on the diet with cholesterol, the level of this parameter was statistically higher in comparison to the group without cholesterol addition. Vanadium treatment in a dose 0.063 mmol/kg of body mass does not influence cholesterol, triglycerides and alanine transaminase activity.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Vanadium, powder, -100 mesh, 99.9% trace metals basis
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Vanadium, pieces, 1-3 mm, 99.9% trace metals basis
Sigma-Aldrich
Vanadium, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Vanadium, rod, diam. 6.2 mm, 99.7% trace metals basis
Vanadium, wire reel, 5m, diameter 0.5mm, as drawn, 99.8%
Vanadium, microfoil, disks, 10mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 10mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 25mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 25mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.2m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 1m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 1m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 1m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 2m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 2m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 2m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 2m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 5m, diameter 0.1mm, as drawn, 99.8%