Merck
  • Inicio
  • Resultados de la búsqueda
  • Determination of deoxynivalenol and deoxynivalenol-3-glucoside in wheat and barley using liquid chromatography coupled to mass spectrometry: on-line clean-up versus conventional sample preparation techniques.
Determination of deoxynivalenol and deoxynivalenol-3-glucoside in wheat and barley using liquid chromatography coupled to mass spectrometry: on-line clean-up versus conventional sample preparation techniques.
Journal of chromatography. A (2014-12-08)
Alexis V Nathanail, Ebru Sarikaya, Marika Jestoi, Michal Godula, Kimmo Peltonen
RESUMEN
In this study, we compared the performance of conventional sample preparation techniques used in mycotoxin analyses against automated on-line sample clean-up for the determination of deoxynivalenol (DON) and its conjugated derivative, deoxynivalenol-3-β-d-glucoside (D3G), in cereal grains. Blank wheat and barley samples were spiked with DON and D3G, extracted with a mixture of acetonitrile:water (84:16, v/v) and processed by one of the following: extract and shoot, MycoSep(®) 227 clean-up columns, MycoSep 227 with an additional acetonitrile elution step and centrifugal filtration, followed by analysis with liquid chromatography tandem mass spectrometry. Based on method performance characteristics and poor recoveries (<30%) obtained for the polar D3G with some techniques, the extract and shoot approach was chosen for the inter-laboratory method comparison study. Thus, the same spiked samples were analysed in parallel by another laboratory with an in-house validated on-line sample clean-up method, utilising TurboFlow™ chromatography coupled to high resolution mass spectrometry. Method validation was performed by determination of specificity, linearity, recovery, intra-day precision and the limits of detection and quantification. Matrix-matched linearity (R(2)>0.985) was established in the range of 100-1600 and 20-320μg/kg for DON and D3G, respectively. Average recoveries (%RSD) were acceptable with both methods for wheat and barley, ranging between 73% and 102% (3-12%) for DON and 72% and 98% (1-10%) for D3G. The benefit of using automated sample clean-up in comparison to extract and shoot is the ability to inject directly pure extracts into the mass spectrometer, offering faster analyses and improved sensitivity with minimum system maintenance.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Deoxynivalenol
Supelco
Deoxynivalenol, reference material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)