• Inicio
  • Resultados de la búsqueda
  • The methanol extract of Euonymus laxiflorus, Rubia lanceolata and Gardenia jasminoides inhibits xanthine oxidase and reduce serum uric acid level in rats.
The methanol extract of Euonymus laxiflorus, Rubia lanceolata and Gardenia jasminoides inhibits xanthine oxidase and reduce serum uric acid level in rats.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-05-23)
Li-Min Liu, Shu-Fen Cheng, Po-Chuen Shieh, Jang-Chang Lee, Jih-Jung Chen, Chi-Tang Ho, Sheng-Chu Kuo, Daih-Huang Kuo, Li-Jiau Huang, Tzong-Der Way
RESUMEN
Chinese herbal medicinal plants, Euonymus laxiflorus (EL), Rubia lanceolata (RL) and Gardenia jasminoides (GJ), have been used wildly to treat arthritis and gout in Taiwan for decades. To understand the beneficial effects of these three plants, their xanthine oxidase (XO) inhibitory activity in vitro and hypouricaemic activity in vivo were investigated. Our results suggested that methanol extracts were better than water extracts for inhibition of XO activity and 2,2-diphenyl-1-picrylhydrazyl (DPPH) radical scavenging activity, except the water extract of GJ, which exhibited the strongest radical scavenging effect. In animal study, the serum urate level was significantly decreased after oral administration of higher dose (0.39g/kg) methanol extract of the mixture of three plants (ERG). In addition, methanol extract of ERG reduced the pain reaction time in the second phase of formalin induced pain. The results provide useful information on the pharmacological activities of these plants for the potential in treating hyperuricemia.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Supelco
Alpha Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Supelco
(±)-α-Tocopherol, analytical standard
Supelco
Gallic acid, certified reference material, TraceCERT®
Sigma-Aldrich
Xanthine, ≥99%
USP
Alpha Tocopherol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
Allopurinol, xanthine oxidase inhibitor
α-Tocopherol, European Pharmacopoeia (EP) Reference Standard
Chlorogenic acid, primary reference standard
Sigma-Aldrich
Xanthine, ≥99.5% (HPLC), purified by recrystallization
Supelco
Allopurinol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(±)-α-Tocopherol, tested according to Ph. Eur.
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
Oxonic acid potassium salt, 97%
Chlorogenic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Xanthine, BioUltra, ≥99%
Allopurinol, European Pharmacopoeia (EP) Reference Standard
USP
Allopurinol, United States Pharmacopeia (USP) Reference Standard