  • Search Results
  • Involvement of TRPV1 and TRPA1 in incisional intraoral and extraoral pain.
Involvement of TRPV1 and TRPA1 in incisional intraoral and extraoral pain.
Journal of dental research (2015-01-13)
K Urata, M Shinoda, K Honda, J Lee, M Maruno, R Ito, N Gionhaku, K Iwata
ABSTRACT
Thermal and mechanical hypersensitivity in the injured region is a common complication. Although it is well known clinically that thermal and mechanical sensitivity of the oral mucosa is different from that of the skin, the mechanisms underlying injured pain of the oral mucosa remain poorly understood. The transient receptor potential (TRP) vanilloid 1 (TRPV1) and TRP ankyrin 1 (TRPA1) in primary afferent neurons are known to contribute to pathological pain. Therefore, we investigated whether TRPV1 and/or TRPA1 contribute to thermal and mechanical hypersensitivity following oral mucosa or whisker pad skin incision. Strong heat and mechanical and cold hypersensitivity was caused in the buccal mucosa and whisker pad skin following incisions. On day 3 after the incisions, the number of TRPV1-immunoreactive (IR) and TRPA1-IR trigeminal ganglion (TG) neurons innervating the buccal mucosa and whisker pad skin was significantly increased, and the number of TRPV1/TRPA1-IR TG neurons innervating whisker pad skin, but not the buccal mucosa, was significantly increased. Administration of the TRPV1 antagonist, SB366791, to the incised site produced a significant suppression of heat hyperalgesia in both the buccal mucosa and whisker pad skin, as well as mechanical allodynia in the whisker pad skin. Administration of the TRPA1 antagonist, HC-030031, to the incised site suppressed mechanical allodynia and cold hyperalgesia in both the buccal mucosa and whisker pad skin, as well as heat hyperalgesia in the whisker pad skin. These findings indicate that altered expressions of TRPV1 and TRPA1 in TG neurons are involved in thermal and mechanical hypersensitivity following the buccal mucosa and whisker pad skin incision. Moreover, diverse changes in the number of TRPV1 and TRPA1 coexpressed TG neurons in whisker pad skin-incised rats may contribute to the intracellular interactions of TRPV1 and TRPA1 associated with whisker pad skin incision, whereas TRPV1 and TRPA1 expression in individual TG neurons is involved in buccal mucosa-incised pain.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
SB-366791, ≥98% (HPLC), powder
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate, 97%
Sigma-Aldrich
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate, BioReagent, suitable for fluorescence, ≥98.0% (TLC)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Sucrose, analytical standard
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material