  • Search Results
  • The antiosteoporotic activity of central-icaritin (CIT) on bone metabolism of ovariectomized rats.
The antiosteoporotic activity of central-icaritin (CIT) on bone metabolism of ovariectomized rats.
Molecules (Basel, Switzerland) (2014-11-19)
Jun Jiang, Jie Li, Xiaobin Jia
ABSTRACT
Central-icaritin (CIT) is a flavonoid aglycone first discovered in our laboratory, which is an isomeric aglycone of icaritin (IT). We wanted to know whether CIT also had anti-osteoporosis activity. In this study, CIT was investigated in an ovariectomized rat (OVX) model. Fifty-six 6-month old female Sprague-Dawley rats were randomly assigned to sham operated group (Sham) and six OVX subgroups (n=8 each). The OVX rats were then subdivided into six groups treated with vehicle (OVX), icaritin (IT, 40 mg/kg body weight/day), estradiol valerate (EV, 100 μg/kg body weight/day) or CIT (10, 20, and 40 mg/kg body weight/day) for 12 weeks, respectively. Then, the serum biochemical parameters, bone mineral density (BMD), bone biomechanical properties, bone microarchitecture, bone immunohistochemistry and related protein and gene expressions were evaluated. In OVX rats, the increases of body weight, HOP, AKP, and TRACP5b levels, and the decreases of uterus wet weight, femurs weight, BMD, serum OPG/RANKL and OCN were significantly inhibited by CIT treatment. Micro-CT analysis results showed that CIT apparently enhanced trabecular bone compared with the OVX group (p<0.05). Total femur BMD and biomechanical strength of tibia were significantly improved (p<0.05) after 12 weeks of CIT administration. In addition, the CIT administration also significantly enhanced the OPG expression, whereas reduced the RANKL expression in femurs according to RT-PCR, western blot assays and immunohistochemical evaluation. CIT had the antiosteoporotic activity, and its antiosteoporotic effects in OVX rats may be stronger than that of IT.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)