Merck
Anginex lipoplexes for delivery of anti-angiogenic siRNA.
International journal of pharmaceutics (2014-06-21)
Afrouz Yousefi, Meriem Bourajjaj, Negar Babae, Paula I van Noort, Roel Q J Schaapveld, Judy R van Beijnum, Arjan W Griffioen, Gert Storm, Raymond M Schiffelers, Enrico Mastrobattista
RESUMEN
Angiogenesis is one of the hallmarks of cancer which renders it an attractive target for therapy of malignancies. Tumor growth suppression can be achieved by inhibiting angiogenesis since it would deprive tumor cells of oxygen and vital nutrients. Activation of endothelial cells of tumor vasculature is the first step in angiogenesis which is mediated by various factors. One of the major triggers in this process is vascular endothelial growth factor (VEGF) which binds to VEGF receptors on endothelial cells of tumor vessels. This induces a series of signaling cascades leading to activation of cellular processes involved in angiogenesis, and therefore down-regulation of VEGF receptor-2 (VEGFR-2) expression seems a viable option to inhibit angiogenesis. In our investigations, this aim has been pursued by using siRNA interfering with the expression of VEGFR-2. Since the discovery of RNA interference (RNAi) as a gene regulation process, successful delivery of small non-coding RNA has presented itself as a major challenge. In the current study, we have characterized a galectin-1 targeted anginex-coupled lipoplex (Angiplex) containing siRNA against the gene of VEGFR-2 as an angiostatic therapeutic. Angiplex particles had a size of approximately 120 nm with a net negative charge and were stable in vitro. These particles were internalized in a specific manner by HUVECs compared to a non-targeted lipoplex system, and their uptake was higher than Lipofectamine 2000. Gene silencing efficiency of Angiplex was shown to be 61%.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
MISSION® esiRNA, targeting human CHN1
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
SAFC
BIS-TRIS
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
SAFC
BIS-TRIS
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Cholesteryl hemisuccinate
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Cholesteryl hemisuccinate tris salt, anionic detergent
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
L-Ascorbic acid, 99%