Merck
Differentiation of Schwann‑like cells from human umbilical cord blood mesenchymal stem cells in vitro.
Molecular medicine reports (2014-11-06)
Yu-Zhou Xiao, Sheng Wang
RESUMEN
The use of artificial nerves for the repair of peripheral nerve defects is restricted by the limited sources of Schwann cells (SCs). Human mesenchymal stem cell (MSC)‑derived Schwann‑like cells are considered an alternative and desirable cell source. The aim of the present study was to establish a method of inducing directional differentiation of human umbilical cord blood (hUCB) MSCs into Schwann‑like cells. Cells isolated from hUCB were cultured in MesenCult complete medium, a specialized culture medium for MSCs, to expand hUCBMSCs. hUCBMSCs were purified by repeated changing of the medium, and they were identified by detection of the specific cell surface markers for MSCs. For differentiation of Schwann‑like cells from hUCBMSCs, the purified cells were sequentially cultured in DMEM/F12 medium with various additives. Differentiated Schwann‑like cells were identified by the detection of SC‑specific markers, including S100b, glial fibrillary acidic protein and P75, by immunocytochemisty, reverse transcription‑polymerase chain reaction and western blotting. The results demonstrated that the majority of the differentiated cells presented classical dipolar and fusiform SC morphology. Notably, a large proportion of these cells expressed the three SC markers. These results suggest that hUCBMSCs can undergo directional differentiation into Schwann‑like cells in vitro and may be an important source of SCs for the treatment of peripheral nerve defects with tissue‑engineered artificial nerves.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Supelco
Forskolin, analytical standard
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Sodium fluoride 0.5 M solution
SAFC
Sodium deoxycholate
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)