  • Search Results
  • Zoledronic acid exerts antitumor effects in NB4 acute promyelocytic leukemia cells by inducing apoptosis and S phase arrest.
Zoledronic acid exerts antitumor effects in NB4 acute promyelocytic leukemia cells by inducing apoptosis and S phase arrest.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2014-10-15)
Shou-Sheng Liu, Xiao-Pai Wang, Xiu-Bo Li, Jia-Yi Liang, Ling-Ling Liu, Ying Lu, Xue-Yun Zhong, Yun-Xian Chen
ABSTRACT
The aim of this study was to investigate the antitumor effect of zoledronic acid (ZOL) in the NB4 human acute promyelocytic leukemia (APL) cell line and explore the potential mechanism of action of this compound. NB4 cells were exposed to various concentrations (0-200μM) of ZOL. Cell viability was measured by MTS assay. The extent of cell apoptosis and distribution of cells in the different phases of the cell cycle were analyzed with flow cytometry. The expression of apoptosis- and cell cycle-related proteins was assayed by Western blot. The combined effect of ZOL and arsenic trioxide (ATO) on the proliferation of NB4 cells was also determined. The results of this study indicate that ZOL inhibits cell proliferation in a time- and dose-dependent fashion and also induces apoptosis and S phase arrest in a dose-dependent manner. The Western blot analysis confirmed the induction of apoptosis and S phase arrest, revealing that the pro-apoptosis proteins Bax, Puma and activated caspase-9 were upregulated and the anti-apoptosis proteins Bcl-2 and Bcl-xL were downregulated. ZOL at a concentration of 50μM synergized with 0.5μM ATO on the growth inhibition of NB4 cells. Taken together, our data indicate that ZOL exerts a potent antitumor effect on NB4 cells by inducing apoptosis and cell cycle arrest, and that ZOL can synergize with the traditional chemotherapy drug ATO.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology