Adipose tissue chromium and vanadium disbalance in high-fat fed Wistar rats.
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS) (2014-09-10)
Alexey A Tinkov, Elizaveta V Popova, Valentina S Polyakova, Olga V Kwan, Anatoly V Skalny, Alexandr A Nikonorov
RESUMEN
The primary objective of the current study is to investigate the relationship between adipose tissue chromium and vanadium content and adipose tissue dysfunction in a model of diet-induced obesity. A total of 26 female Wistar rats were fed either standard or high-fat diet (31.6% of fat from total caloric content) for 3 months. High-fat-feeding resulted in 21 and 33% decrease in adipose tissue chromium and vanadium content, respectively. No change was seen in hair chromium or vanadium levels. Statistical analysis revealed a significant inverse correlation of adipose tissue Cr and V with animal morphometric parameters and adipocyte size. Significant inverse dependence was observed between adipose tissue Cr and V and serum leptin and proinflammatory cytokines' levels. At the same time, adipose tissue Cr and V levels were characterized by positive correlation between serum adiponectin and adiponectin/leptin ratio. Adipose tissue Cr and V were inversely correlated (p<0.05) with insulin and homeostatic model assessment insulin resistance index (HOMA-IR) levels. Cr and V concentrations were not correlated with serum glucose in either high-fat fed or control rats; however, both serum glucose and HOMA-IR levels were significantly higher in high-fat fed, compared to control, rats. The results allow to hypothesize that impairment of adipose tissue Cr and V content plays a certain role in the development of adipose tissue endocrine dysfunction in obesity.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Vanadium, powder, -100 mesh, 99.9% trace metals basis
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Vanadium, pieces, 1-3 mm, 99.9% trace metals basis
Sigma-Aldrich
Vanadium, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Vanadium, rod, diam. 6.2 mm, 99.7% trace metals basis
Vanadium, wire reel, 5m, diameter 0.5mm, as drawn, 99.8%
Vanadium, microfoil, disks, 10mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 10mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 25mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 25mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.2m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 1m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 1m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 1m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 2m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 2m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 2m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 2m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 5m, diameter 0.1mm, as drawn, 99.8%