Merck
  • Inicio
  • Resultados de la búsqueda
  • In vitro assessment of the multifunctional bioactive potential of Alaska pollock skin collagen following simulated gastrointestinal digestion.
In vitro assessment of the multifunctional bioactive potential of Alaska pollock skin collagen following simulated gastrointestinal digestion.
Journal of the science of food and agriculture (2014-08-02)
Lidong Guo, Pádraigín A Harnedy, Li Zhang, Bafang Li, Zhaohui Zhang, Hu Hou, Xue Zhao, Richard J FitzGerald
RESUMEN
Dietary mineral deficiency, hypertension and diabetes have become serious human health problems. Dietary approaches are increasingly being investigated to address these issues. Identification of food-derived biological peptides has become an important approach to control such diseases. Peptides generated from aquatic byproducts have been shown to possess biological activities. Significantly higher copper-chelating activity was observed on simulated hydrolysis of intact collagen. The collagen hydrolysate generated in the gastric stage exhibited moderate angiotensin-converting enzyme (ACE)-inhibitory activity with an IC50 value of 2.92 ± 0.22 mg mL(-1), which significantly decreased to 0.49 ± 0.02 mg mL(-1) after intestinal digestion. The dipeptidyl peptidase (DPP) IV-inhibitory potency of the collagen hydrolysate generated directly following simulated gastrointestinal digestion (SGID) (IC50 2.59 ± 0.04 mg mL(-1)) was significantly lower than that of the collagen tryptic hydrolysate (CTH) (IC50 1.53 ± 0.01 mg mL(-1)). The antioxidant activities of collagen and CTH using the ferric-reducing antioxidant power (FRAP) assay were 0.87 ± 0.10 and 1.27 ± 0.03 µmol Trolox equivalent (TE) g(-1) respectively after SGID. This study identifies collagen as a good and inexpensive substrate for the generation of biologically active peptides with potential applications as functional ingredients in the management of chronic illness and mineral deficiency problems.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Supelco
Trifluoroacetic acid, analytical standard
Supelco
Acetonitrile, analytical standard
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Coumarin, primary reference standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Ile-Pro-Ile, ≥97% (HPLC)