Elevated expression of pleiotrophin in lymphocytic leukemia CD19+ B cells.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica (2014-04-05)
Chun-Xian Du, Lan Wang, Yan Li, Wei Xiao, Qin-Lian Guo, Fei Chen, Xin-Ti Tan
RESUMEN
Pleiotrophin (PTN) has been demonstrated to be strongly expressed in many fetal tissues, but seldom in healthy adult tissues. While PTN has been reported to be expressed in many types of tumors as well as at high serum concentrations in patients with many types of cancer, to date, there has been no report that PTN is expressed in leukemia, especially in lymphocytic leukemia. We isolated the CD19(+) subset of B cells from peripheral blood from healthy adults, B-cell acute lymphocytic leukemia (B-ALL) patients, and B-cell chronic lymphocytic leukemia (B-CLL) patients and examined these cells for PTN mRNA and protein expression. We used immunocytochemistry, western blotting, and enzyme-linked immunosorbent assay to show that PTN protein is highly expressed in CD19(+) B cells from B-ALL and B-CLL patients, but barely expressed in B cells from healthy adults. We also examined PTN expression at the nucleic acid level using reverse transcription polymerase chain reaction (RT-PCR) and northern blotting and detected a high levels of PTN transcripts in the CD19(+) B cells from both groups of leukemia patients, but very few in the CD19(+) B cells from the healthy controls. Interestingly, the quantity of the PTN transcripts correlated with the severity of disease. Moreover, suppression of PTN activity with an anti-PTN antibody promoted apoptosis of cells from leukemia patients and cell lines SMS-SB and JVM-2. This effect of the anti-PTN antibody suggests that PTN may be a new target for the treatment of lymphocytic leukemia.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pleiotrophin human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Pleiotrophin human, ≥97% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture
Redes sociales
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Investigación. Desarrollo. Producción.
Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos
© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.
Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.