Oxysterols synergize with statins by inhibiting SREBP-2 in ovarian cancer cells.
Gynecologic oncology (2014-08-20)
Cinzia Casella, Daniel H Miller, Kerry Lynch, Alexander S Brodsky
RESUMEN
Determine mechanisms responsible for enhanced statin efficacy in a novel statin combination we name STOX (STatin-OXysterol). Ovarian cancer cell lines were treated with combinations of statins and oxysterols. Cell viability was determined by a modified MTT assay. Apoptosis was evaluated by immunoblotting of PARP and DAPI-mediated visualization of apoptotic nuclei. STOX effects on the expression of genes of the mevalonate pathway were assessed by real-time qPCR and immunoblotting. siRNA-mediated gene silencing was used to test the involvement of oxysterol-mediated repression of SREBP-2 in STOX synergy. The impact of statin-mediated inhibition of protein prenylation and on cholesterol homeostasis was evaluated. Oxysterols dramatically enhance cytotoxicity of statins in ovarian cancer cells through increased apoptosis. Decreased expression of SREBP-2 down-regulates the mevalonate pathway and prevents the active statin-induced sterol feedback, enhancing statin toxicity. Comparison of two ovarian cancer cell lines reveals two distinct mechanisms of statin induced toxicity, namely, dependence on protein geranylgeranylation and/or perturbation of cellular cholesterol levels. We provide evidence of statins' mechanisms of cytotoxicity in different ovarian cancer cells and discovered a new approach to significantly enhance the anti-tumor activity of statins. These observations provide a potential new path to improve statins as a treatment against ovarian cancer with obtainable dosages.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 5 M
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)