Merck
Deficiency of the two-pore-domain potassium channel TREK-1 promotes hyperoxia-induced lung injury.
Critical care medicine (2014-08-16)
Andreas Schwingshackl, Bin Teng, Patrudu Makena, Manik Ghosh, Scott E Sinclair, Charlean Luellen, Louisa Balasz, Cynthia Rovnaghi, Robert M Bryan, Eric E Lloyd, Elizabeth Fitzpatrick, Jordy S Saravia, Stephania A Cormier, Christopher M Waters
RESUMEN
We previously reported the expression of the two-pore-domain K channel TREK-1 in lung epithelial cells and proposed a role for this channel in the regulation of alveolar epithelial cytokine secretion. In this study, we focused on investigating the role of TREK-1 in vivo in the development of hyperoxia-induced lung injury. Laboratory animal experiments. University research laboratory. Wild-type and TREK-1-deficient mice. Mice were anesthetized and exposed to 1) room air, no mechanical ventilation, 2) 95% hyperoxia for 24 hours, and 3) 95% hyperoxia for 24 hours followed by mechanical ventilation for 4 hours. Hyperoxia exposure accentuated lung injury in TREK-1-deficient mice but not controls, resulting in increase in lung injury scores, bronchoalveolar lavage fluid cell numbers, and cellular apoptosis and a decrease in quasi-static lung compliance. Exposure to a combination of hyperoxia and injurious mechanical ventilation resulted in further morphological lung damage and increased lung injury scores and bronchoalveolar lavage fluid cell numbers in control but not TREK-1-deficient mice. At baseline and after hyperoxia exposure, bronchoalveolar lavage cytokine levels were unchanged in TREK-1-deficient mice compared with controls. Exposure to hyperoxia and mechanical ventilation resulted in an increase in bronchoalveolar lavage interleukin-6, monocyte chemotactic protein-1, and tumor necrosis factor-α levels in both mouse types, but the increase in interleukin-6 and monocyte chemotactic protein-1 levels was less prominent in TREK-1-deficient mice than in controls. Lung tissue macrophage inflammatory protein-2, keratinocyte-derived cytokine, and interleukin-1β gene expression was not altered by hyperoxia in TREK-1-deficient mice compared with controls. Furthermore, we show for the first time TREK-1 expression on alveolar macrophages and unimpaired tumor necrosis factor-α secretion from TREK-1-deficient macrophages. TREK-1 deficiency resulted in increased sensitivity of lungs to hyperoxia, but this effect is less prominent if overwhelming injury is induced by the combination of hyperoxia and injurious mechanical ventilation. TREK-1 may constitute a new potential target for the development of novel treatment strategies against hyperoxia-induced lung injury.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone GG-7, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone HP-6017, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Rat IgG2a antibody produced in mouse, clone R2A-2, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Human IgG2 (Fc specific) antibody produced in mouse, clone HP-6002, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Human IgG4 (pFc′ specific) antibody produced in mouse, clone HP-6023, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Human IgG (Fab specific) antibody produced in mouse, clone GG-6, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Rabbit IgG (γ-chain specific) antibody produced in mouse, clone RG-96, ascites fluid
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Biotin antibody, Mouse monoclonal, clone HP-6017, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Goat/Sheep IgG−FITC antibody, Mouse monoclonal, clone GT-34, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG (γ-chain specific)−Biotin antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Goat/Sheep IgG−Peroxidase antibody, Mouse monoclonal, clone GT-34, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG, Native antibody, Mouse monoclonal, clone RabT-50, purified from hybridoma cell culture
Sigma-Aldrich
Rat Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Rat Tumor Necrosis Factor α ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Anti-Human IgG (Fc specific)−FITC antibody, Mouse monoclonal, clone HP-6017, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Goat/Sheep IgG−Biotin antibody, Mouse monoclonal, clone GT-34, purified from hybridoma cell culture
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC), suitable for fluorescence
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Monoclonal Anti-Goat/Sheep IgG antibody produced in mouse, clone GT-34, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Human IgG (γ-chain specific) antibody produced in mouse, clone GG-5, ascites fluid
Sigma-Aldrich
Anti-Rabbit IgG (γ-chain specific)–Peroxidase antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Rabbit IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in mouse, clone RG-96, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Rabbit IgG, Native−Peroxidase antibody, Mouse monoclonal, clone RabT-50, purified from hybridoma cell culture
SAFC
BIS-TRIS
Sigma-Aldrich
Mouse Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant