Merck
First report of a de novo mutation at SLC20A2 in a patient with brain calcification.
Journal of molecular neuroscience : MN (2014-06-28)
J B Ferreira, L Pimentel, M P Keasey, R R Lemos, L M Santos, M F Oliveira, S Santos, N Jensen, K Teixeira, L Pedersen, C R Rocha, M R Dias da Silva, J R M Oliveira
RESUMEN
Primary familial brain calcification (PFBC) is identified by mineralization of the basal ganglia and other brain regions in the absence of known causes. The condition is often inherited in an autosomal dominant pattern and can manifest itself clinically with neuropsychiatric symptoms such as Parkinsonism, headaches, psychosis, and mood swings. Mutations in the SLC20A2 gene account for ~40% of inherited cases, and this gene encodes an inorganic phosphate transporter (PiT-2), a transmembrane protein associated with Pi homeostasis. The p.Y386X mutation in SLC20A2 was identified in a patient who presented migraines, brain calcification, and mild but chronic hypovitaminosis D. SLC20A2 c.1158C > G single-nucleotide heterozygous mutation results in a premature stop codon and a putative truncated protein of 385 amino acids. Proband parents do not present the mutation, which is also not present in major public SNP databases, suggesting a de novo sporadic trait. This study describes for the first time a de novo SLC20A2 mutation in a PFBC patient with migraine and mild hypovitaminosis D. This data further reinforces the pathogenic role of SLC20A2 mutations as causal factors in PFBC physiopathology.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, deionized
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, tested according to Ph. Eur.
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O