Merck
  • Inicio
  • Resultados de la búsqueda
  • A quantitative LC-MS/MS method for determination of SP-141, a novel pyrido[b]indole anticancer agent, and its application to a mouse PK study.
A quantitative LC-MS/MS method for determination of SP-141, a novel pyrido[b]indole anticancer agent, and its application to a mouse PK study.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-09-10)
Subhasree Nag, Jiang-Jiang Qin, Shivaputra Patil, Hemantkumar Deokar, John K Buolamwini, Wei Wang, Ruiwen Zhang
RESUMEN
In the present study, a specific and sensitive liquid chromatography-triple quadrupole mass spectrometry method was developed and validated for the determination of SP-141, a novel pyrido[b]indole anticancer agent. After a liquid-liquid extraction with n-hexane-dichloromethane-2-propanol (20:10:1, v/v/v) mixture, the analyte was separated on a Kinetex C18 column (50×2.1mm, 2.6μm) with mobile phases comprising of water (0.1% formic acid, v/v) and acetonitrile (0.1% formic acid, v/v) at a flow rate of 0.4mL/min. The test compound (SP-141) and the internal standard (SP-157) were analyzed in the multiple reaction-monitoring mode using the mass transitions m/z 325.1 → 282.0. The method was linear in the concentration range of 0.648-162ng/mL with coefficients of determination (R(2)) of 0.999 in mouse plasma. The lower limit of quantification was 0.648ng/mL. The intra- and inter-day assay precisions (coefficient of variation, %CV) were less than 4.2% and accuracies (relative error, %RE) ranged from -6.1% to 2.1%. The extraction recoveries were between 97.1 and 103.1% and the relative matrix effect was minimal. In addition, SP-141 was found to be stable in the plasma after three freeze-thaw cycles, at 37°C and 4°C for 24h, and at -80°C for 4 weeks. It was also stable in the stock solution at room temperature for 24h and after preparation in the autosampler for 36h. The validated method was successfully applied to an initial pharmacokinetic study of SP-141 in CD-1 mice following intraperitoneal and intravenous administrations.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Methylene chloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)