Merck
Reactive oxygen species regulate the quiescence of CD34-positive cells derived from human embryonic stem cells.
Cardiovascular research (2014-04-22)
Sun-Hwa Song, Kyungjong Kim, Jeong Joo Park, Kyung Hoon Min, Wonhee Suh
RESUMEN
Reactive oxygen species (ROS) are involved in a wide range of cellular processes. However, few studies have examined the generation and function of ROS in human embryonic vascular development. In this study, the sources of ROS and their roles in the vascular differentiation of human embryonic stem cells (hESCs) were investigated. During vascular differentiation of hESCs, CD34(+) cells had quiescence-related gene expression profiles and a large fraction of these cells were in G0 phase. In addition, levels of ROS, which were primarily generated through NOX4, were substantially higher in hESC-derived CD34(+) cells than in hESC-derived CD34(-) cells. To determine whether excess levels of ROS induce quiescence of hESC-derived CD34(+) cells, ROS levels were moderately reduced using selenium to enhance antioxidant activities of thioredoxin reductase and glutathione peroxidase. In comparison to untreated CD34(+) cells, selenium-treated CD34(+) cells exhibited changes in gene expression that favoured cell cycle progression, and had a greater proliferation and a smaller fraction of cells in G0 phase. Thus, selenium treatment increased the number of hESC-derived CD34(+) cells, thereby enhancing the efficiency with which hESCs differentiated into vascular endothelial and smooth muscle cells. This study reveals that NOX4 produces ROS in CD34(+) cells during vascular differentiation of hESCs, and shows that modulation of ROS levels using antioxidants such as selenium may be a novel approach to increase the vascular differentiation efficiency of hESCs.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Pyronin Y, certified by the Biological Stain Commission, Dye content 50 %
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Pyronin Y, for microscopy (Bot., Fl., Hist.)
Sigma-Aldrich
Pyronin Y, for NA electrophoresis
Sigma-Aldrich
Allopurinol, xanthine oxidase inhibitor
Allopurinol, European Pharmacopoeia (EP) Reference Standard
Supelco
Allopurinol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Allopurinol, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Mercaptosuccinic acid, ReagentPlus®, ≥99.0% (HPLC)
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
Mercaptosuccinic acid, 97%
Sigma-Aldrich
4′-Hydroxy-3′-methoxyacetophenone, 98%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Acetovanillone, ≥98%, FG
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)