Gene expression profiling of sesaminol triglucoside and its tetrahydrofuranoid metabolites in primary rat hepatocytes.
International journal of food sciences and nutrition (2014-08-27)
Kuo-Ching Jan, Binghuei-Barry Yang, Tristan C Liu
RESUMEN
Sesaminol triglucoside is a major lignin in sesame meal and has a methylenedioxyphenyl group and multiple functions in vivo. As a tetrahydrofurofuran type lignan, sesaminol triglucoside is metabolized to mammalian lignans. This investigation studies the effect of sesaminol triglucoside and its tetrahydrofuranoid metabolites (sesaminol, 2-episesaminol, hydroxymethyl sesaminol-tetrahydrofuran, enterolactone, and enterodiol) on gene expression in primary rat hepatocytes using a DNA microarray. Sesame lignans significantly affected the expression of xenobiotic-induced transcripts of cytochrome P450, solute carrier (SLC), and ATP-binding cassette (ABC) transporters. Changes in gene expression were generally greater in response to metabolites with methylenedioxyphenyl moieties (sesaminol triglucoside, sesaminol, and 2-episesaminol) than to the tetrahydrofuranoid metabolites (hydroxymethyl sesaminol-tetrahydrofuran, enterolactone, and enterodiol). Tetrahydrofuran lignans, such as sesaminol triglucoside, sesamin, hydroxymethyl sesaminol-tetrahydrofuran, and sesaminol changed the expression of ABC transporters.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
SAFC
HEPES
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Serine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
Enterolactone, ~95% (HPLC)
Sigma-Aldrich
Enterodiol, ≥95.0% (HPLC)
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material