Isolation and epithelial co-culture of mouse renal peritubular endothelial cells.
BMC cell biology (2014-12-01)
Ye Zhao, Hong Zhao, Yun Zhang, Tania Tsatralis, Qi Cao, Ya Wang, Yiping Wang, Yuan Min Wang, Steve I Alexander, David C Harris, Guoping Zheng
RESUMEN
Endothelial-mesenchymal transition (EndoMT) has been shown to be a major source of myofibroblasts, contributing to kidney fibrosis. However, in vitro study of endothelial cells often relies on culture of isolated primary endothelial cells due to the unavailability of endothelial cell lines. Our recent study suggested that peritubular endothelial cells could contribute to kidney fibrosis through EndoMT. Therefore, successful isolation and culture of mouse peritubular endothelial cells could provide a new platform for studying kidney fibrosis. This study describes an immunomagnetic separation method for the isolation of mouse renal peritubular endothelial cells using anti-CD146 MicroBeads, followed by co-culture with mouse renal proximal tubular epithelial cells to maintain endothelial phenotype. Flow cytometry showed that after isolation and two days of culture, about 95% of cells were positive for endothelial-specific marker CD146. The percentage of other cells, including dendritic cells (CD11c) and macrophages (F4/80), was less than 1%. Maintenance of endothelial cell phenotype required vascular endothelial growth factor (VEGF) and co-culture with mouse proximal tubular epithelial cells. In this study, we established a method for the isolation of mouse renal peritubular endothelial cells by using immunomagnetic separation with anti-CD146 MicroBeads, followed by co-culture with mouse renal proximal tubular epithelial cells to maintain phenotype.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, for residue analysis
Supelco
Ethanol, standard for GC
Supelco
Methanol, analytical standard
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade