Merck
  • Inicio
  • Resultados de la búsqueda
  • Occupational exposure to lead decreases macular, choroidal, and retinal nerve fiber layer thickness in industrial battery workers.
Occupational exposure to lead decreases macular, choroidal, and retinal nerve fiber layer thickness in industrial battery workers.
Current eye research (2014-02-08)
Metin Ekinci, Erdinç Ceylan, H Hüseyin Cağatay, Sadullah Keleş, Hasan Altınkaynak, Baki Kartal, Yaran Koban, Nergiz Hüseyinoğlu
RESUMEN
To evaluate the toxic effects of chronic lead (Pb) exposure on retinal nerve fiber layer thickness (RNFLT), macular thickness (MT) and choroidal thickness (CT) in battery industry workers. Male factory workers (n = 50) and healthy non-employees (n = 20) participated in the study. Group 1 (n = 22) comprised lead workers; Group 2 (n = 16), box makers; Group 3 (n = 12), assistant personnel; and Group 4 (n = 20), healthy non-employees. All participants were given Best-Corrected Visual Acuity (BCVA) tests, full ophthalmologic examinations, and Spectral Domain Optical Coherence Tomography (SD-OCT) of the optic nerve head, RNFL, macula, and choroid. Blood lead levels (BLL) in venous blood samples were determined. The duration of exposure to lead varied, and is expressed in months. Average (mean ± SD) ages of participants were 27.55 ± 4.00, 28.69 ± 3.48, 32.00 ± 3.38, and 29.85 ± 5.48 yrs (Groups 1-4, respectively). BLL were 46.2 ± 2.32, 29.31 ± 3.30, 16.9 ± 1.9, and 2.85 ± 0.98 µg/dL (Groups 1-4, respectively). Durations of exposure to lead were 43.86 ± 10.81, 42.81 ± 4.86, and 49.42 ± 6.14 mo (Groups 1-3, respectively). OCT averages for RNFL were 101.68 ± 5.32, 119.50 ± 13.47, 127.67 ± 8.92, and 130.9 ± 6.63 μm (Groups 1-4, respectively). Although RNFLTs of Group 1 were significantly less than those of for Groups 2-4, and, RNFLTs of Groups 2 and 3 were significantly less than that of Group 4, there were no significant differences between Group 2 and 3. MTs were 94.50 ± 6.78, 105.63 ± 5.43, 111.50 ± 6.74 and 147.95 ± 6.67 μm, (Groups 1-4, respectively). CTs were 176.41 ± 15.39, 222.19 ± 17.79, 239.17 ± 15.64, and 251.50 ± 10.98 μm (Groups 1-4, respectively). Both MTs and CTs displayed significant differences among the four groups. Ocular changes in individuals who are chronically exposed to lead include decrease in RNFT, MT, and CT, and thus these parameters should be evaluated during ophthalmologic examination of individuals working in lead-based industries.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Trimethylphenylammonium hydroxide solution, ~25% in H2O (1.68 M)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
Trimethylphenylammonium hydroxide solution, ~0.5 M (CH3)3N(OH)C6H5 in methanol, for GC derivatization, LiChropur