Merck
  • Inicio
  • Resultados de la búsqueda
  • The effects of vasoactive peptide urocortin 2 on hemodynamics in spontaneous hypertensive rat and the role of L-type calcium channel and CRFR2.
The effects of vasoactive peptide urocortin 2 on hemodynamics in spontaneous hypertensive rat and the role of L-type calcium channel and CRFR2.
Pharmacological reports : PR (2015-02-26)
Chunna Liu, Xinyu Liu, Jing Yang, Yan Duan, Hongyue Yao, Fenghua Li, Xia Zhang
RESUMEN
Urocortin (UCN) is a newly identified vascular-active peptide that has been shown to reverse cardiovascular remodeling and improve left ventricular (LV) function. The effects and mechanism of urocortin 2 (UCN2) in vivo on the electrical remodeling of left ventricle and the hemodynamics of hypertensive objectives have not been investigated. UCN2 (1 μg/kg/d, 3.5 μg/kg/d or 7 μg/kg/d) was intravenously injected for 2 weeks and its effects on hemodynamics in spontaneously hypertensive rats (SHRs) observed. The whole-cell patch clamp technique was used to explore the effects of UCN2 on the electrical remodeling of left ventricular cardiomyocytes. The flow cytometry method was used to determine the content of fluorescence calcium in myocardium. UCN2 improved the systolic and diastolic function of SHRs as demonstrated by decreased left ventricular systolic pressure (LVSP), left ventricular end diastolic pressure (LVEDP), increased +dp/dtmax and -dp/dtmax and decreased cAMP level. UCN2 inhibited the opening of L-type calcium channel and decreased the calcium channel current of cardiomyocytes. In addition, UCN2 also decreased the contents of fluorescence calcium in SHR myocardium. However, astressin2-B (AST-2B), the antagonist of corticotropin-releasing factor receptor 2 (CRFR2), could reverse the inhibitory effects of UCN2 on calcium channel. UCN2 can modulate electrical remodeling of the myocardium and hemodynamics in an experimental model of SHR via inhibition of L-type calcium channel and CRFR2 in cardiomyocytes.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, deionized
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, tested according to Ph. Eur.
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Water, ACS reagent, for ultratrace analysis