The hyperglycemic byproduct methylglyoxal impairs anticoagulant activity through covalent adduction of antithrombin III.
Thrombosis research (2014-10-14)
Richard Jacobson, Nicholas Mignemi, Kristie Rose, Lynda O'Rear, Suryakala Sarilla, Heidi E Hamm, Joey V Barnett, Ingrid M Verhamme, Jonathan Schoenecker
RESUMEN
The blood coagulation system is a tightly regulated balance of procoagulant and anticoagulant factors, disruption of which can cause clinical complications. Diabetics are known to have a hypercoagulable phenotype, along with increased circulating levels of methylglyoxal (MGO) and decreased activity of the anticoagulant plasma protein antithrombin III (ATIII). MGO has been shown to inhibit ATIII activity in vitro, however the mechanism of inhibition is incompletely understood. As such, we designed this study to investigate the kinetics and mechanism of MGO-mediated ATIII inhibition. MGO-mediated ATIII inhibition was confirmed using inverse experiments detecting activity of the ATIII targets thrombin and factor Xa. Fluorogenic assays were performed in both PBS and plasma after incubation of ATIII with MGO, at molar ratios comparable to those observed in the plasma of diabetic patients. LC-coupled tandem mass spectrometry was utilized to investigate the exact mechanism of MGO-mediated ATIII inhibition. MGO concentration-dependently attenuated inhibition of thrombin and factor Xa by ATIII in PBS-based assays, both in the presence and absence of heparin. In addition, MGO concentration-dependently inhibited ATIII activity in a plasma-based system, to the level of plasma completely deficient in ATIII, again both in the presence and absence of heparin. Results from LC-MS/MS experiments revealed that MGO covalently adducts the active site Arg 393 of ATIII through two distinct glyoxalation mechanisms. We posit that active site adduction is the mechanism of MGO-mediated inhibition of ATIII, and thus contributes to the underlying pathophysiology of the diabetic hypercoagulable state and complications thereof.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water
Sigma-Aldrich
Methylglyoxal solution, ~40% in H2O
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%