Merck
Gender-specific differences in the in situ cardiac function of endotoxemic rats detected by pressure-volume catheter.
Shock (Augusta, Ga.) (2014-07-09)
Ronald P M Gonçalves, Karla L Guarido, Jamil Assreuy, J Eduardo da Silva-Santos
RESUMEN
The aim of the present study was to investigate the existence of gender-related differences in the profile of changes that occur in cardiac functionality during endotoxic shock. For this, both male and female Wistar rats received a single injection of lipopolysaccharide (LPS; 10 mg/kg, i.p.) at 6 h (LPS 6-h group) or 24 h (LPS 24-h group) before the induction of anesthesia and insertion of a pressure-volume catheter using the closed-chest method. Control animals received sterile saline. Hemodynamic parameters were recorded under basal conditions and during the peak of the pressor effect of phenylephrine (30 nmol/kg i.v.). Body temperature, hematologic parameters, blood glucose, and diuresis were also evaluated. There were unremarkable differences between male and female rats in the general aspects of sepsis evaluated in our study. Both male and female rats from the LPS 6-h group presented hypotension, depressed left ventricular ejection fraction, decreased stroke work, reduced dP/dtmax (maximal rate of left ventricle pressure change), P@dP/dtmax (pressure value at the maximum dP/dtmax), dP/dtmin (minimal rate of left ventricle pressure change), and preload-recruitable stroke work indices, as well as increased end-systolic volume. Nevertheless, only male rats from the LPS 24-h group still presented decreased stroke work and reduced dP/dtmax, P@dP/dtmax, and preload-recruitable stroke work indices. The end-systolic volume presented slight changes during the pressor effects of phenylephrine in all groups of male rats, as well as in females from the control and LPS 6-h groups, but it was significantly increased in females from the LPS 24-h group. These findings suggest that after induction of endotoxic shock female rats may recover the inotropic cardiac function earlier than males, as well as present improved adaptation of their left ventricle to the pressure-loading effects of phenylephrine.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®